Assay Details
| Gene Symbol: Sh3bgrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509193.2 | XP_003509241.1 | 3 - 4 | 519 | 67 | |
| GenBank mRNA | JI872180.1 | - | 3 - 4 | 539 | 67 | |
| JP047301.1 | - | 3 - 4 | 539 | 67 | ||
More Information
Set Membership: |
|

Sh3bgrl
Genomic Map