Assay Details
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511221.2 | XP_003511269.1 | 6 - 7 | 791 | 97 | |
XM_007651749.1 | XP_007649939.1 | 6 - 7 | 752 | 97 | ||
XM_007651750.1 | XP_007649940.1 | 6 - 7 | 1142 | 97 | ||
XM_007651751.1 | XP_007649941.1 | 6 - 7 | 691 | 97 | ||
XM_007651752.1 | XP_007649942.1 | 6 - 7 | 1190 | 97 | ||
GenBank mRNA | JI876222.1 | - | 1 - 2 | 175 | 97 | |
JP038279.1 | - | 1 - 2 | 175 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
