Assay Details
| Gene Symbol: Clpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512695.2 | XP_003512743.1 | 11 - 12 | 1388 | 56 | |
| XM_007653104.1 | XP_007651294.1 | 12 - 13 | 1533 | 56 | ||
| XM_007653105.1 | XP_007651295.1 | 12 - 13 | 1478 | 56 | ||
| GenBank mRNA | JI876173.1 | - | 8 - 9 | 790 | 56 | |
More Information
Set Membership: |
|

Clpb
Genomic Map