Assay Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650703.1 | XP_007648893.1 | 2 - 3 | 352 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab3b| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650703.1 | XP_007648893.1 | 2 - 3 | 352 | 75 | |