Assay Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653562.1 | XP_007651752.1 | 3 - 4 | 550 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb10| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653562.1 | XP_007651752.1 | 3 - 4 | 550 | 52 | |
Set Membership: |
|