Assay Details
| Gene Symbol: Znf496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643952.1 | XP_007642142.1 | 7 - 8 | 1123 | 69 | |
| XM_007643953.1 | XP_007642143.1 | 7 - 8 | 1051 | 69 | ||
| XM_007643954.1 | XP_007642144.1 | 7 - 8 | 1050 | 69 | ||
| XM_007643955.1 | XP_007642145.1 | 7 - 8 | 1076 | 69 | ||
More Information
Set Membership: |
|

Znf496
Genomic Map