Assay Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642041.1 | XP_007640231.1 | 6 - 7 | 617 | 64 | |
| XM_007642044.1 | XP_007640234.1 | 6 - 7 | 616 | 64 | ||
| XM_007642049.1 | XP_007640239.1 | 6 - 7 | 678 | 64 | ||
| GenBank mRNA | JI891287.1 | - | 5 - 6 | 601 | 64 | |
More Information
Set Membership: |
|

Il10rb
Genomic Map