Assay Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643418.1 | XP_007641608.1 | 3 - 4 | 660 | 69 | |
| XM_007643419.1 | XP_007641609.1 | 4 - 5 | 789 | 69 | ||
More Information
Set Membership: |
|

Esrrb
Genomic Map