Assay Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641748.1 | XP_007639938.1 | 3 - 4 | 1075 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb7c| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641748.1 | XP_007639938.1 | 3 - 4 | 1075 | 80 | |
Set Membership: |
|