Assay Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496071.2 | XP_003496119.1 | 4 - 5 | 648 | 64 | |
| GenBank mRNA | JI872990.1 | - | 1 - 2 | 64 | ||
More Information
Set Membership: |
|

Emg1
Genomic Map