Assay Details
| Gene Symbol: Itgb3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497609.2 | XP_003497657.2 | 3 - 4 | 254 | 77 | |
| XM_007614538.1 | XP_007612728.1 | 3 - 4 | 332 | 77 | ||
| GenBank mRNA | JI894061.1 | - | 1 - 2 | 134 | 77 | |
More Information
Set Membership: |
|

Itgb3bp
Genomic Map