Assay Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501145.2 | XP_003501193.1 | 9 - 10 | 1131 | 59 | |
| GenBank mRNA | JI871817.1 | - | 8 - 9 | 1091 | 59 | |
More Information
Set Membership: |
|

Nfs1
Genomic Map