Assay Details
| Gene Symbol: Cideb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502087.2 | XP_003502135.2 | 3 - 4 | 355 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cideb| Gene Symbol: Cideb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502087.2 | XP_003502135.2 | 3 - 4 | 355 | 58 | |
Set Membership: |
|