Assay Details
| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504613.2 | XP_003504661.1 | 4 - 5 | 559 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi35| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504613.2 | XP_003504661.1 | 4 - 5 | 559 | 60 | |
Set Membership: |
|