Assay Details
| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI879110.1 | - | 6 - 7 | 669 | 74 | |
| JP055538.1 | - | 6 - 7 | 669 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754902| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI879110.1 | - | 6 - 7 | 669 | 74 | |
| JP055538.1 | - | 6 - 7 | 669 | 74 | ||