Assay Details
| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI893495.1 | - | 2 - 3 | 221 | 61 | |
| JP056179.1 | - | 2 - 3 | 221 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754902| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI893495.1 | - | 2 - 3 | 221 | 61 | |
| JP056179.1 | - | 2 - 3 | 221 | 61 | ||
Set Membership: |
|