Assay Details
| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI879110.1 | - | 3 - 4 | 386 | 66 | |
| JP055538.1 | - | 3 - 4 | 386 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754902| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI879110.1 | - | 3 - 4 | 386 | 66 | |
| JP055538.1 | - | 3 - 4 | 386 | 66 | ||
Set Membership: |
|