Assay Details
| Gene Symbol: Nxpe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509322.1 | XP_003509370.1 | 2 - 3 | 126 | 93 | |
| XM_007649906.1 | XP_007648096.1 | 1 - 2 | 144 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxpe2| Gene Symbol: Nxpe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509322.1 | XP_003509370.1 | 2 - 3 | 126 | 93 | |
| XM_007649906.1 | XP_007648096.1 | 1 - 2 | 144 | 93 | ||