Assay Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007613594.1 | XP_007611784.1 | 6 - 7 | 409 | 65 | |
| GenBank mRNA | JI876845.1 | - | 6 - 7 | 675 | 65 | |
More Information
Set Membership: |
|

Ube2h
Genomic Map