Assay Details
| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654385.1 | XP_007652575.1 | 7 - 8 | 731 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754902| Gene Symbol: LOC100754902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654385.1 | XP_007652575.1 | 7 - 8 | 731 | 73 | |
Set Membership: |
|