Assay Details
| Gene Symbol: Ccdc104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503979.2 | XP_003504027.1 | 4 - 5 | 587 | 90 | |
| XM_007644505.1 | XP_007642695.1 | 4 - 5 | 587 | 90 | ||
| GenBank mRNA | JI870683.1 | - | 4 - 5 | 588 | 90 | |
| JP049858.1 | - | 4 - 5 | 588 | 90 | ||
More Information
Set Membership: |
|

Ccdc104
Genomic Map