Assay Details
| Gene Symbol: Tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506662.2 | XP_003506710.1 | 4 - 5 | 1977 | 64 | |
| GenBank mRNA | JI887490.1 | - | 4 - 5 | 870 | 64 | |
More Information
Set Membership: |
|

Tdgf1
Genomic Map