Assay Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514083.2 | XP_003514131.1 | 4 - 5 | 503 | 72 | |
| GenBank mRNA | JI877770.1 | - | 4 - 5 | 651 | 72 | |
More Information
Set Membership: |
|

Rras
Genomic Map