Assay Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642208.1 | XP_007640398.1 | 2 - 3 | 2462 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bricd5| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642208.1 | XP_007640398.1 | 2 - 3 | 2462 | 62 | |
Set Membership: |
|