Assay Details
| Gene Symbol: Ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498650.2 | XP_003498698.1 | 1 - 2 | 203 | 80 | |
| XM_007631125.1 | XP_007629315.1 | 1 - 2 | 203 | 80 | ||
| XM_007631140.1 | XP_007629330.1 | 1 - 2 | 201 | 80 | ||
| GenBank mRNA | JI865370.1 | - | 1 - 2 | 204 | 80 | |
More Information
Set Membership: |
|

Ensa
Genomic Map