Assay Details
| Gene Symbol: Csrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498861.2 | XP_003498909.1 | 4 - 5 | 540 | 66 | |
| GenBank mRNA | JI887373.1 | - | 4 - 5 | 471 | 66 | |
| JP039534.1 | - | 4 - 5 | 471 | 66 | ||
More Information
Set Membership: |
|

Csrp1
Genomic Map