Assay Details
| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501132.2 | XP_003501180.1 | 3 - 4 | 380 | 62 | |
| XM_007641829.1 | XP_007640019.1 | 3 - 4 | 402 | 62 | ||
| GenBank mRNA | JI889007.1 | - | 3 - 4 | 342 | 62 | |
| JP042328.1 | - | 3 - 4 | 342 | 62 | ||
More Information
Set Membership: |
|

Gss
Genomic Map