Assay Details
| Gene Symbol: Pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504748.2 | XP_003504796.1 | 1 - 2 | 421 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgam2| Gene Symbol: Pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504748.2 | XP_003504796.1 | 1 - 2 | 421 | 74 | |
Set Membership: |
|