Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI872885.1 | - | 613 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccbl2| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI872885.1 | - | 613 | 119 | ||