Assay Details
| Gene Symbol: Rpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641235.1 | XP_007639425.1 | 4 - 5 | 513 | 59 | |
| GenBank mRNA | JP040887.1 | - | 1 - 2 | 134 | 59 | |
More Information
Set Membership: |
|

Rpl4
Genomic Map