Assay Details
| Gene Symbol: Akt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645768.1 | XP_007643958.1 | 9 - 10 | 1240 | 65 | |
| GenBank mRNA | JI878273.1 | - | 2 - 3 | 464 | 65 | |
| JP060248.1 | - | 2 - 3 | 464 | 65 | ||
More Information
Set Membership: |
|

Akt2
Genomic Map