Assay Details
| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652811.1 | XP_007651001.1 | 1 - 2 | 715 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a1| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652811.1 | XP_007651001.1 | 1 - 2 | 715 | 79 | |
Set Membership: |
|