Assay Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007628159.1 | XP_007626349.1 | 3 - 4 | 458 | 67 | |
| GenBank mRNA | JI882455.1 | - | 3 - 4 | 458 | 67 | |
More Information
Set Membership: |
|

Diablo
Genomic Map