Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 82 - 83 | 19903 | 78 | |
| XM_007645556.1 | XP_007643746.1 | 82 - 83 | 19903 | 78 | ||
| XM_007645557.1 | XP_007643747.1 | 81 - 82 | 19840 | 78 | ||
| XM_007645558.1 | XP_007643748.1 | 77 - 78 | 13807 | 78 | ||
| XM_007645559.1 | XP_007643749.1 | 77 - 78 | 13807 | 78 | ||
| XM_007645560.1 | XP_007643750.1 | 76 - 77 | 13744 | 78 | ||
| XM_007645561.1 | XP_007643751.1 | 26 - 27 | 8531 | 78 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map