Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650651.1 | XP_007648841.1 | 3 - 4 | 347 | 63 | |
| GenBank mRNA | JI872885.1 | - | 1251 | 63 | ||
More Information
Set Membership: |
|

Ccbl2
Genomic Map