Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498467.2 | XP_003498515.1 | 5 - 6 | 575 | 73 | |
| GenBank mRNA | JI865415.1 | - | 6 - 7 | 74 | ||
More Information
Set Membership: |
|

Fam216a
Genomic Map