Assay Details
| Gene Symbol: Ss18l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500690.2 | XP_003500738.1 | 1 - 2 | 164 | 78 | |
| GenBank mRNA | JI872429.1 | - | 1 - 2 | 145 | 78 | |
| JP047275.1 | - | 1 - 2 | 145 | 78 | ||
More Information
Set Membership: |
|

Ss18l2
Genomic Map