Assay Details
| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501918.2 | XP_003501966.1 | 5 - 6 | 863 | 71 | |
| GenBank mRNA | JI880442.1 | - | 5 - 6 | 780 | 71 | |
| JP042154.1 | - | 5 - 6 | 780 | 71 | ||
More Information
Set Membership: |
|

Dnase2
Genomic Map