Assay Details
| Gene Symbol: Ces1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503623.1 | XP_003503671.1 | 7 - 8 | 898 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ces1| Gene Symbol: Ces1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503623.1 | XP_003503671.1 | 7 - 8 | 898 | 85 | |
Set Membership: |
|