Assay Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504248.2 | XP_003504296.1 | 2 - 3 | 296 | 71 | |
| GenBank mRNA | JI880281.1 | - | 2 - 3 | 168 | 71 | |
More Information
Set Membership: |
|

Elp6
Genomic Map