Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647585.1 | XP_007645775.1 | 5 - 6 | 458 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2cr11| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647585.1 | XP_007645775.1 | 5 - 6 | 458 | 89 | |
Set Membership: |
|