Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654641.1 | XP_007652831.1 | 3 - 4 | 409 | 76 | |
| GenBank mRNA | JI885508.1 | - | 1 - 2 | 169 | 76 | |
More Information
Set Membership: |
|

Ubfd1
Genomic Map