Assay Details
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640107.1 | XP_007638297.1 | 5 - 6 | 717 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
