Assay Details
| Gene Symbol: LOC103163567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650494.1 | XP_007648684.1 | 5 - 6 | 552 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103163567| Gene Symbol: LOC103163567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650494.1 | XP_007648684.1 | 5 - 6 | 552 | 79 | |
Set Membership: |
|