Assay Details
| Gene Symbol: LOC100766662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497636.2 | XP_003497684.1 | 3 - 4 | 660 | 62 | |
| GenBank mRNA | JI863427.1 | - | 3 - 4 | 552 | 62 | |
More Information
Set Membership: |
|

LOC100766662
Genomic Map