Assay Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498038.2 | XP_003498086.2 | 9 - 10 | 1086 | 69 | |
| XM_007621795.1 | XP_007619985.1 | 10 - 11 | 1389 | 69 | ||
| XM_007621802.1 | XP_007619992.1 | 9 - 10 | 1238 | 69 | ||
| XM_007621811.1 | XP_007620001.1 | 10 - 11 | 1135 | 69 | ||
| XM_007621817.1 | XP_007620007.1 | 10 - 11 | 1237 | 69 | ||
| XM_007621831.1 | XP_007620021.1 | 9 - 10 | 832 | 69 | ||
| GenBank mRNA | JI893020.1 | - | 3 - 4 | 298 | 69 | |
More Information
Set Membership: |
|

Adal
Genomic Map