Assay Details
| Gene Symbol: Znf775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501331.2 | XP_003501379.2 | 2 - 3 | 236 | 65 | |
| XM_007641974.1 | XP_007640164.1 | 3 - 4 | 279 | 65 | ||
| XM_007641975.1 | XP_007640165.1 | 2 - 3 | 174 | 65 | ||
| XM_007641976.1 | XP_007640166.1 | 2 - 3 | 114 | 65 | ||
More Information
Set Membership: |
|

Znf775
Genomic Map