Assay Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503612.2 | XP_003503660.1 | 5 - 6 | 685 | 72 | |
| XM_007644203.1 | XP_007642393.1 | 5 - 6 | 682 | 72 | ||
| GenBank mRNA | JI895938.1 | - | 5 - 6 | 671 | 72 | |
| JP041760.1 | - | 5 - 6 | 671 | 72 | ||
More Information
Set Membership: |
|

Herpud1
Genomic Map