Assay Details
| Gene Symbol: Rad9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509938.2 | XP_003509986.1 | 7 - 8 | 1842 | 69 | |
| XM_007650596.1 | XP_007648786.1 | 5 - 6 | 557 | 69 | ||
| GenBank mRNA | JI872465.1 | - | 7 - 8 | 834 | 69 | |
More Information
Set Membership: |
|

Rad9a
Genomic Map