Assay Details
| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511698.2 | XP_003511746.1 | 1 - 2 | 255 | 75 | |
| GenBank mRNA | JI890569.1 | - | 1 - 2 | 255 | 75 | |
| JP062444.1 | - | 1 - 2 | 255 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mesdc2| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511698.2 | XP_003511746.1 | 1 - 2 | 255 | 75 | |
| GenBank mRNA | JI890569.1 | - | 1 - 2 | 255 | 75 | |
| JP062444.1 | - | 1 - 2 | 255 | 75 | ||